missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Parainfluenza Type-2 Protein
Recombinant Protein
£99.15 - £2205.00
Specifications
Name | Influenza Parainfluenza Type-2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942079
|
enQuireBio™
QP12960-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962079
|
enQuireBio™
QP12960-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952079
|
enQuireBio™
QP12960-1MG |
1 mg |
£2205.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza Parainfluenza Type-2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Parainfluenza type-2 solution contains STE, 0.02% sodium azide (NaN3). |