missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Parainfluenza Type-2 Protein
Recombinant Protein
Brand: enQuireBio™ QP12960-1mg
This item is not returnable.
View return policy
Specifications
Influenza Parainfluenza Type-2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Parainfluenza type-2 solution contains STE, 0.02% sodium azide (NaN3). |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |