missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YWHAE Protein
A cDNA sequence encoding the YWHAE was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13985-5ug
Additional Details : Weight : 0.01000kg
Specifications
7531 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDDREDLVYQ AKLAEQAERY DEMVESMKKV AGMDVELTVE ERNLLSVAYK NVIGARRASW RIISSIEQKE ENKGGEDKLK MIREYRQMVE TELKLICCDI LDVLDKHLIP AANTGESKVF YYKMKGDYHR YLAEFATGND RKEAAENSLV AYKAASDIAM TELPPTHPIR LGLALNFSVF YYEILNSPDR ACRLAKAAFD DAIAELDTLS EESYKDSTLI MQLLRDNLTL WTSDMQGDGE EQNKEALQDV EDENQ | |
Greater than 90.0% as determined by SDS-PAGE. |
YWHAE Protein | |
Research Use Only | |
YWHAE | |
Human | |
Untagged | |
The YWHAE solution (1 mg/1ml) contains phosphate buffered Saline (pH 7.4), 1mM DTT and 10% glycerol. |