missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YWHAE Protein
A cDNA sequence encoding the YWHAE was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Gene ID (Entrez) | 7531 |
---|---|
Name | YWHAE Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | YWHAE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975319
|
enQuireBio™
QP13985-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965319
|
enQuireBio™
QP13985-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955319
|
enQuireBio™
QP13985-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7531 | |
Research Use Only | |
YWHAE | |
Human | |
Untagged | |
The YWHAE solution (1 mg/1ml) contains phosphate buffered Saline (pH 7.4), 1mM DTT and 10% glycerol. |
YWHAE Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDDREDLVYQ AKLAEQAERY DEMVESMKKV AGMDVELTVE ERNLLSVAYK NVIGARRASW RIISSIEQKE ENKGGEDKLK MIREYRQMVE TELKLICCDI LDVLDKHLIP AANTGESKVF YYKMKGDYHR YLAEFATGND RKEAAENSLV AYKAASDIAM TELPPTHPIR LGLALNFSVF YYEILNSPDR ACRLAKAAFD DAIAELDTLS EESYKDSTLI MQLLRDNLTL WTSDMQGDGE EQNKEALQDV EDENQ | |
Greater than 90.0% as determined by SDS-PAGE. |