missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human WIF-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£395.24
Specifications
Gene ID (Entrez) | 11197 |
---|---|
Molecular Weight (g/mol) | 39.8 kDa |
Name | WIF-1 |
Quantity | 50μg |
Source | Mouse myeloma cell line,NS0-derived human WIF-1 protein Gly29-Trp379,with a C-terminal 10-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18146660
|
R&D Systems™
1341-WF-050 |
Lyophilized from a 0.2μm filtered solution in Acetic Acid, NaCl, PEG3350 and CHAPS with BSA as a carrier protein. |
£395.24
50µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18187390
|
R&D Systems™
1341-WF-050/CF |
Lyophilized from a 0.2μm filtered solution in Acetic Acid, NaCl, PEG3350 and CHAPS. |
£395.24
50µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human WIF-1 Protein is derived from NS0. The Recombinant Human WIF-1 Protein has been validated for the following applications: Bioactivity.Specifications
11197 | |
WIF-1 | |
Mouse myeloma cell line,NS0-derived human WIF-1 protein Gly29-Trp379,with a C-terminal 10-His tag | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
39.8 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |