missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human WIF-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1341-WF-050/CF
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human WIF-1 Protein is derived from NS0. The Recombinant Human WIF-1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetic Acid, NaCl, PEG3350 and CHAPS. | |
39.8 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
11197 | |
WIF-1 | |
Mouse myeloma cell line,NS0-derived human WIF-1 protein Gly29-Trp379,with a C-terminal 10-His tag | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |