missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGF-165 Isoform Protein
Recombinant Protein
£80.80 - £4075.00
Specifications
Accession Number | NP_001165097.1 |
---|---|
Name | VEGF-165 Isoform Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | VEGFA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965689
|
enQuireBio™
QP5210-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945689
|
enQuireBio™
QP5210-10UG |
10 μg |
£198.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935689
|
enQuireBio™
QP5210-100UG |
100 μg |
£535.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975689
|
enQuireBio™
QP5210-500UG |
500 μg |
£2495.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955689
|
enQuireBio™
QP5210-1MG |
1 mg |
£4075.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001165097.1 | |
Research Use Only | |
VEGFA | |
Recombinant Protein | |
E. coli | |
MAPMAEGGGQ NHHEVVKFMD VYQRSYCHPI ETLVDIFQEY PDEIEYIFKP SCVPLMRCGG CCNDEGLECV PTEESNITMQ IMRIKPHQGQ HIGEMSFLQH NKCECRPKKD RARQENPCGP CSERRKHLFV QDPQTCKCSC KNTDSRCKAR QLELNERTCR CDKPRR | |
Greater than 95% as determined by SDS-PAGE. |
VEGF-165 Isoform Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
HUVEC proliferation is significantly induced by less than 10 ng/ml of Human VEGF-165 Isoform, or activity of at least 1.0 x 10^5 units/mg. Typical ED50 values are less than 5 ng/ml. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |