missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VEGF-165 Isoform Protein

Recombinant Protein

Brand:  enQuireBio™ QP5210-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955689

  • £4075.00 / 1mg
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001165097.1
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
HUVEC proliferation is significantly induced by less than 10 ng/ml of Human VEGF-165 Isoform, or activity of at least 1.0 x 10^5 units/mg. Typical ED50 values are less than 5 ng/ml.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
VEGF-165 Isoform Protein
Research Use Only
VEGFA
Recombinant Protein
E. coli
MAPMAEGGGQ NHHEVVKFMD VYQRSYCHPI ETLVDIFQEY PDEIEYIFKP SCVPLMRCGG CCNDEGLECV PTEESNITMQ IMRIKPHQGQ HIGEMSFLQH NKCECRPKKD RARQENPCGP CSERRKHLFV QDPQTCKCSC KNTDSRCKAR QLELNERTCR CDKPRR
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers