missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human VCAM-1/CD106 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£348.80 - £902.78
Specifications
Accession Number | P19320-1 |
---|---|
Formulation | Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. |
Gene ID (Entrez) | 7412 |
Molecular Weight (g/mol) | 74.1 kDa |
Name | VCAM-1/CD106 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18390388
|
R&D Systems™
ADP5-050 |
£348.80
50µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18158180
|
R&D Systems™
ADP5-200 |
200μg |
£902.78
200µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human VCAM-1/CD106 Protein is derived from CHO. The Recombinant Human VCAM-1/CD106 Protein has been validated for the following applications: Bioactivity.Specifications
P19320-1 | |
7412 | |
VCAM-1/CD106 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. | |
74.1 kDa | |
Chinese Hamster Ovary cell line,CHO-derived human VCAM-1/CD106 protein Phe25-Glu698 (Arg322Gly) | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |