missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human VCAM-1/CD106 Protein
Click to view available options
Quantity:
200 μg
50 μg
Unit Size:
200µg
50µg
Description
The Recombinant Human VCAM-1/CD106 Protein is derived from CHO. The Recombinant Human VCAM-1/CD106 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P19320-1 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. |
| Gene ID (Entrez) | 7412 |
| Molecular Weight (g/mol) | 74.1 kDa |
| Name | VCAM-1/CD106 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human VCAM-1/CD106 protein Phe25-Glu698 (Arg322Gly) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction