missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vacuolar fusion protein MON1 homolog A Protein
A DNA sequence encoding the Homo sapiens (Human) Vacuolar fusion protein MON1 homolog A, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001135973.1 |
---|---|
Gene ID (Entrez) | 84315 |
Name | Vacuolar fusion protein MON1 homolog A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050871
|
enQuireBio™
QP7799-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070871
|
enQuireBio™
QP7799-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060871
|
enQuireBio™
QP7799-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080871
|
enQuireBio™
QP7799-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090871
|
enQuireBio™
QP7799-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000881
|
enQuireBio™
QP7799-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001135973.1 | |
Vacuolar fusion protein MON1 homolog A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
84315 | |
Research Use Only | |
MON1A | |
Recombinant Protein | |
E. coli | |
DGYKVVFVRR SPLVLVAVAR TRQSAQELAQ ELLYIYYQIL SLLTGAQLSH IFQQKQNYDL RRLLSGSERI TDNLLQLMAR DPSFLMGAAR CLPLAAAVRD TVSASLQQAR ARSLVFSILL ARNQLVALVR RKDQFLHPID LHLLFNLISS SSSFREGEAW TPVCLPKFNA AGFFHAHISY LEPDTDLCLL LVSTDREDFF AVSDCRRRFQ ERLRKRGAHL ALREALRTPY YSVAQVGIPD LRHFLYKSKS SGLFTSPEIE APYTSEEEQE RLLGLYQYLH SRAHNASRPL KTIYYTGPNE NLLAWVTGAF ELYMCYSPLG TKASAVSAIH KLMRWIRKEE DRLFILTPLT Y | |
Greater than 90% as determined by SDS-PAGE. |