missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Uncharacterized protein C1orf106 Protein

A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C1orf106, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9786-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008721

  • £467.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001136041.1
Uncharacterized protein C1orf106 Protein
Research Use Only
C1orf106
Recombinant Protein
Yeast
MLQMPKLNEI PPGRAGRREA RGEGRWPGQT GPEAARLEWR AQGQAGGARA PWDSWGSSRL PTQPGPGWSR CPPSLLCALS FQKSTMESKD EVSDTDSGII LQSGPDSPVS PMKELTHAVH KQQRALEARL EACLEELRRL CLREAELTGT LPAEYPLKPG EKAPKVRRRI GAAYKLDDWA LHREDPLSSL ERQLALQLQI TEAARRLCLE ENLSRQARRQ RKHSMLQEEK KLQELQRCLV ERRRNSEPPP AAALPLGREL SASDDSSLSD GLLLEEEESQ VPKPPPESPA PPSRPLPPQT LEGLQPTGPE AGSPERAPVQ NSPWKETSLD HPYEKPRKSS EPWSESSSPA TTPQDGPSAS SLWLLEPASY HVVPIRGVPG QWQGRTSAPA TPEIQGRRGQ SQSLRVDSFR AGPEGRGRSA FPRRRPTHYT VTVPDSCFPA TKPPLPHAAC HSCSEDSGSD VSSISHPTSP GSSSPDISFL QPLSPPKTHR HRGAWVPAGS RELVAHHPKL LLPPGYFPAG RYVVVAESPL PPGEWELRRA APGPAYEEEG TPLRYQRLVP SRSRIVRTPS LKDSPAGR
Greater than 90% as determined by SDS-PAGE.
55765
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers