missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C1orf106 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C1orf106, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_001136041.1 |
---|---|
Gene ID (Entrez) | 55765 |
Name | Uncharacterized protein C1orf106 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008721
|
enQuireBio™
QP9786-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048721
|
enQuireBio™
QP9786-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018721
|
enQuireBio™
QP9786-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038721
|
enQuireBio™
QP9786-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058721
|
enQuireBio™
QP9786-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028721
|
enQuireBio™
QP9786-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001136041.1 | |
Uncharacterized protein C1orf106 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55765 | |
Research Use Only | |
C1orf106 | |
Recombinant Protein | |
Yeast | |
MLQMPKLNEI PPGRAGRREA RGEGRWPGQT GPEAARLEWR AQGQAGGARA PWDSWGSSRL PTQPGPGWSR CPPSLLCALS FQKSTMESKD EVSDTDSGII LQSGPDSPVS PMKELTHAVH KQQRALEARL EACLEELRRL CLREAELTGT LPAEYPLKPG EKAPKVRRRI GAAYKLDDWA LHREDPLSSL ERQLALQLQI TEAARRLCLE ENLSRQARRQ RKHSMLQEEK KLQELQRCLV ERRRNSEPPP AAALPLGREL SASDDSSLSD GLLLEEEESQ VPKPPPESPA PPSRPLPPQT LEGLQPTGPE AGSPERAPVQ NSPWKETSLD HPYEKPRKSS EPWSESSSPA TTPQDGPSAS SLWLLEPASY HVVPIRGVPG QWQGRTSAPA TPEIQGRRGQ SQSLRVDSFR AGPEGRGRSA FPRRRPTHYT VTVPDSCFPA TKPPLPHAAC HSCSEDSGSD VSSISHPTSP GSSSPDISFL QPLSPPKTHR HRGAWVPAGS RELVAHHPKL LLPPGYFPAG RYVVVAESPL PPGEWELRRA APGPAYEEEG TPLRYQRLVP SRSRIVRTPS LKDSPAGR | |
Greater than 90% as determined by SDS-PAGE. |