missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TPT1 Protein
A cDNA sequence encoding the TPT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13800-25ug
Additional Details : Weight : 0.01000kg
Specifications
TPT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMMI IYRDLISHDE MFSDIYKIRE IADGLCLEVE GKMVSRTEGN IDDSLIGGNA SAEGPEGEGT ESTVITGVDI VMNHHLQETS FTKEAYKKYI KDYMKSIKGK LEEQRPERVK PFMTGAAEQI KHILANFKNY QFFIGENMNP DGMVALLDYR EDGVTPYMIF FKDGLEMEKC | |
Greater than 90% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TPT1 solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol. |