missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TPT1 Protein
A cDNA sequence encoding the TPT1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2490.00
Specifications
Name | TPT1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954749
|
enQuireBio™
QP13800-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944749
|
enQuireBio™
QP13800-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934749
|
enQuireBio™
QP13800-1MG |
1 mg |
£2490.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TPT1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TPT1 solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMMI IYRDLISHDE MFSDIYKIRE IADGLCLEVE GKMVSRTEGN IDDSLIGGNA SAEGPEGEGT ESTVITGVDI VMNHHLQETS FTKEAYKKYI KDYMKSIKGK LEEQRPERVK PFMTGAAEQI KHILANFKNY QFFIGENMNP DGMVALLDYR EDGVTPYMIF FKDGLEMEKC | |
Greater than 90% as determined by SDS-PAGE. |