missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TPT1 Protein

A cDNA sequence encoding the TPT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13800-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934749

  • £2490.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TPT1 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMMI IYRDLISHDE MFSDIYKIRE IADGLCLEVE GKMVSRTEGN IDDSLIGGNA SAEGPEGEGT ESTVITGVDI VMNHHLQETS FTKEAYKKYI KDYMKSIKGK LEEQRPERVK PFMTGAAEQI KHILANFKNY QFFIGENMNP DGMVALLDYR EDGVTPYMIF FKDGLEMEKC
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TPT1 solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers