missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFA Protein
A cDNA sequence encoding the TGFA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10875-1mg
Additional Details : Weight : 0.01000kg
Specifications
TGFA Protein | |
Research Use Only | |
The ED50, as measured in a proliferation assay using mouse Balb/3T3 cells, is less than 0.4ng/ml. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
VVSHFNDCPD SHTQFCFHGT CRFLVQEDKP ACVCHSGYVG ARCEHADLLA | |
Greater than 97.0% as determined by SDS-PAGE. |