missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFA Protein
A cDNA sequence encoding the TGFA was constructed and used to recombinantly synthesize the protein.
£99.15 - £1095.00
Specifications
Name | TGFA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50, as measured in a proliferation assay using mouse Balb/3T3 cells, is less than 0.4ng/ml. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945438
|
enQuireBio™
QP10875-20UG |
20 μg |
£99.15
20µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925438
|
enQuireBio™
QP10875-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935438
|
enQuireBio™
QP10875-1MG |
1 mg |
£1095.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TGFA Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
VVSHFNDCPD SHTQFCFHGT CRFLVQEDKP ACVCHSGYVG ARCEHADLLA | |
Greater than 97.0% as determined by SDS-PAGE. |
Research Use Only | |
The ED50, as measured in a proliferation assay using mouse Balb/3T3 cells, is less than 0.4ng/ml. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |