missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TEF Protein

A cDNA sequence encoding the TEF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13717-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15974479

  • £99.15 / 2µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

7008
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSDAGGG KKPPVDPQAG PGPGPGRAAG ERGLSGSFPL VLKKLMENPP REARLDKEKG KEKLEEDEAA AASTMAVSAS LMPPIWDKTI PYDGESFHLE YMDLDEFLLE NGIPASPTHL AHNLLLPVAE LEGKESASSS TASPPSSSTA IFQPSETVSS TESSLEKERE TPSPIDPNCV EVDVNFNPDP ADLVLSSVPG GELFNPRKHK FAEEDLKPQP MIKKAKKVFV PDEQKDEKYW TRRKKNNVAA KRSRDARRLK ENQITIRAAF LEKENTALRT EVAELRKEVG KCKTIVSKYE TKYGPL
Greater than 85% as determined by SDS-PAGE.
TEF Protein
Research Use Only
TEF
Human
His
The TEF solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers