missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TEF Protein
A cDNA sequence encoding the TEF was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 7008 |
---|---|
Name | TEF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TEF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974479
|
enQuireBio™
QP13717-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964479
|
enQuireBio™
QP13717-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954479
|
enQuireBio™
QP13717-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7008 | |
Research Use Only | |
TEF | |
Human | |
His | |
The TEF solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
TEF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSDAGGG KKPPVDPQAG PGPGPGRAAG ERGLSGSFPL VLKKLMENPP REARLDKEKG KEKLEEDEAA AASTMAVSAS LMPPIWDKTI PYDGESFHLE YMDLDEFLLE NGIPASPTHL AHNLLLPVAE LEGKESASSS TASPPSSSTA IFQPSETVSS TESSLEKERE TPSPIDPNCV EVDVNFNPDP ADLVLSSVPG GELFNPRKHK FAEEDLKPQP MIKKAKKVFV PDEQKDEKYW TRRKKNNVAA KRSRDARRLK ENQITIRAAF LEKENTALRT EVAELRKEVG KCKTIVSKYE TKYGPL | |
Greater than 85% as determined by SDS-PAGE. |