missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Syncytin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Syncytin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8241-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001124397.1 | |
Syncytin-1 Protein | |
Research Use Only | |
ERVW-1 | |
Recombinant Protein | |
E. coli | |
APPPCRCMTS SSPYQEFLWR MQRPGNIDAP SYRSLSKGTP TFTAHTHMPR NCYHSATLCM HANTHYWTGK MINPSCPGGL GVTVCWTYFT QTGMSDGGGV QDQAREKHVK EVISQLTRVH GTSSPYKGLD LSKLHETLRT HTRLVSLFNT TLTGLHEVSA QNPTNCWICL PLNFRPYVSI PVPEQWNNFS TEINTTSVLV GPLVSNLEIT HTSNLTCVKF SNTTYTTNSQ CIRWVTPPTQ IVCLPSGIFF VCGTSAYRCL NGSSESMCFL SFLVPPMTIY TEQDLYSYVI SKPRNKRVPI LPFVIGAGVL GALGTGIGGI TTSTQFYYKL SQELNGDMER VADSLVTLQD QLNSLAAVVL QNRRALDLLT AERGGTCLFL GEECCYYVNQ SGIVTEKVKE IRDRIQRRAE ELRNTGPWGL LSQ | |
Greater than 90% as determined by SDS-PAGE. |
30816 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |