missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Syncytin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Syncytin-1, was expressed in the hosts and tags indicated.
£228.00 - £2480.00
Specifications
Accession Number | NP_001124397.1 |
---|---|
Gene ID (Entrez) | 30816 |
Name | Syncytin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013501
|
enQuireBio™
QP8241-EC-10UG |
10 μg |
£228.00
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003501
|
enQuireBio™
QP8241-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023501
|
enQuireBio™
QP8241-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033501
|
enQuireBio™
QP8241-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043501
|
enQuireBio™
QP8241-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053501
|
enQuireBio™
QP8241-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001124397.1 | |
Syncytin-1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
30816 | |
Research Use Only | |
ERVW-1 | |
Recombinant Protein | |
E. coli | |
APPPCRCMTS SSPYQEFLWR MQRPGNIDAP SYRSLSKGTP TFTAHTHMPR NCYHSATLCM HANTHYWTGK MINPSCPGGL GVTVCWTYFT QTGMSDGGGV QDQAREKHVK EVISQLTRVH GTSSPYKGLD LSKLHETLRT HTRLVSLFNT TLTGLHEVSA QNPTNCWICL PLNFRPYVSI PVPEQWNNFS TEINTTSVLV GPLVSNLEIT HTSNLTCVKF SNTTYTTNSQ CIRWVTPPTQ IVCLPSGIFF VCGTSAYRCL NGSSESMCFL SFLVPPMTIY TEQDLYSYVI SKPRNKRVPI LPFVIGAGVL GALGTGIGGI TTSTQFYYKL SQELNGDMER VADSLVTLQD QLNSLAAVVL QNRRALDLLT AERGGTCLFL GEECCYYVNQ SGIVTEKVKE IRDRIQRRAE ELRNTGPWGL LSQ | |
Greater than 90% as determined by SDS-PAGE. |