missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STAC Protein
A cDNA sequence encoding the STAC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13605-1mg
Additional Details : Weight : 0.01000kg
Specifications
STAC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMIPPSSP REDGVDGLPK EAVGAEQPPS PASTSSQESK LQKLKRSLSF KTKSLRSKSA DNFFQRTNSE DMKLQAHMVA EISPSSSPLP APGSLTSTPA RAGLHPGGKA HAFQEYIFKK PTFCDVCNHM IVGTNAKHGL RCKACKMSIH HKCTDGLAPQ RCMGKLPKGF RRYYSSPLLI HEQFGCIKEV MPIACGNKVD PVYETLRFGT SLAQRTKKGS SGSGSDSPHR TSTSDLVEVP EEANGPGGGY DLRKRSNSVF TYPENGTDDF RDPAKNINHQ GSLSKDPLQM NTYVALYKFV PQENEDLEMR PGDIITLLED SNEDWWKGKI QDRIGFFPAN FVQRLQQNEK IFRCVRTFIG CKEQGQITLK ENQICVSSEE EQDGFIRVLS GKKKGLIPLD VLENI | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STAC protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |