missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STAC Protein
A cDNA sequence encoding the STAC was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | STAC Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964119
|
enQuireBio™
QP13605-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954119
|
enQuireBio™
QP13605-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944119
|
enQuireBio™
QP13605-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
STAC Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STAC protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMIPPSSP REDGVDGLPK EAVGAEQPPS PASTSSQESK LQKLKRSLSF KTKSLRSKSA DNFFQRTNSE DMKLQAHMVA EISPSSSPLP APGSLTSTPA RAGLHPGGKA HAFQEYIFKK PTFCDVCNHM IVGTNAKHGL RCKACKMSIH HKCTDGLAPQ RCMGKLPKGF RRYYSSPLLI HEQFGCIKEV MPIACGNKVD PVYETLRFGT SLAQRTKKGS SGSGSDSPHR TSTSDLVEVP EEANGPGGGY DLRKRSNSVF TYPENGTDDF RDPAKNINHQ GSLSKDPLQM NTYVALYKFV PQENEDLEMR PGDIITLLED SNEDWWKGKI QDRIGFFPAN FVQRLQQNEK IFRCVRTFIG CKEQGQITLK ENQICVSSEE EQDGFIRVLS GKKKGLIPLD VLENI | |
Greater than 85.0% as determined by SDS-PAGE. |