missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SRP54 Protein
A cDNA sequence encoding the SRP54 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13591-5ug
Additional Details : Weight : 0.01000kg
Specifications
SRP54 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MVLADLGRKI TSALRSLSNA TIINEEVLNA MLKEVCTALL EADVNIKLVK QLRENVKSAI DLEEMASGLN KRKMIQHAVF KELVKLVDPG VKAWTPTKGK QNVIMFVGLQ GSGKTTTCSK LAYYYQRKGW KTCLICADTF RAGAFDQLKQ NATKARIPFY GSYTEMDPVI IASEGVEKFK NENFEIIIVD TSGRHKQEDS LFEEMLQVAN AIQPDNIVYV MDASIGQACE AQAKAFKDKV DVASVIVTKL DGHAKGGGAL SAVAATKSPI IFIGTGEHID DFEPFKTQPF ISKLLGMGDI EGLIDKVNEL KLDDNEALIE KLKHGQFTLR DMYEQFQNIM KMGPFSQILG MIPGFGTDFM SKGNEQESMA RLKKLMTIMD SMNDQELDST DGAKVFSKQP GRIQRVARGS GVSTRDVQEL LTQYTKFAQM VKKMGGIKGL FKGGDMSKNV SQSQMAKLNQ QMAKMMDPRV LHHMGGMAGL QSMMRQFQQG AAGNMKGMMG FNNM | |
Greater than 80% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SRP54 is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol. |