missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SRP54 Protein
A cDNA sequence encoding the SRP54 was constructed and used to recombinantly synthesize the protein.
£99.15 - £4295.00
Specifications
Name | SRP54 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944079
|
enQuireBio™
QP13591-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934079
|
enQuireBio™
QP13591-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924079
|
enQuireBio™
QP13591-1MG |
1 mg |
£4295.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SRP54 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SRP54 is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MVLADLGRKI TSALRSLSNA TIINEEVLNA MLKEVCTALL EADVNIKLVK QLRENVKSAI DLEEMASGLN KRKMIQHAVF KELVKLVDPG VKAWTPTKGK QNVIMFVGLQ GSGKTTTCSK LAYYYQRKGW KTCLICADTF RAGAFDQLKQ NATKARIPFY GSYTEMDPVI IASEGVEKFK NENFEIIIVD TSGRHKQEDS LFEEMLQVAN AIQPDNIVYV MDASIGQACE AQAKAFKDKV DVASVIVTKL DGHAKGGGAL SAVAATKSPI IFIGTGEHID DFEPFKTQPF ISKLLGMGDI EGLIDKVNEL KLDDNEALIE KLKHGQFTLR DMYEQFQNIM KMGPFSQILG MIPGFGTDFM SKGNEQESMA RLKKLMTIMD SMNDQELDST DGAKVFSKQP GRIQRVARGS GVSTRDVQEL LTQYTKFAQM VKKMGGIKGL FKGGDMSKNV SQSQMAKLNQ QMAKMMDPRV LHHMGGMAGL QSMMRQFQQG AAGNMKGMMG FNNM | |
Greater than 80% as determined by SDS-PAGE. |