missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SRP54 Protein

A cDNA sequence encoding the SRP54 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13591-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924079

  • £4295.00 / 1mg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SRP54 Protein
Research Use Only
Recombinant Protein
Baculovirus
MVLADLGRKI TSALRSLSNA TIINEEVLNA MLKEVCTALL EADVNIKLVK QLRENVKSAI DLEEMASGLN KRKMIQHAVF KELVKLVDPG VKAWTPTKGK QNVIMFVGLQ GSGKTTTCSK LAYYYQRKGW KTCLICADTF RAGAFDQLKQ NATKARIPFY GSYTEMDPVI IASEGVEKFK NENFEIIIVD TSGRHKQEDS LFEEMLQVAN AIQPDNIVYV MDASIGQACE AQAKAFKDKV DVASVIVTKL DGHAKGGGAL SAVAATKSPI IFIGTGEHID DFEPFKTQPF ISKLLGMGDI EGLIDKVNEL KLDDNEALIE KLKHGQFTLR DMYEQFQNIM KMGPFSQILG MIPGFGTDFM SKGNEQESMA RLKKLMTIMD SMNDQELDST DGAKVFSKQP GRIQRVARGS GVSTRDVQEL LTQYTKFAQM VKKMGGIKGL FKGGDMSKNV SQSQMAKLNQ QMAKMMDPRV LHHMGGMAGL QSMMRQFQQG AAGNMKGMMG FNNM
Greater than 80% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SRP54 is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers