missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNIP1 Protein

A cDNA sequence encoding the SNIP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13539-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15923899

  • £2975.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SNIP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MRWRLYPFKN DEVLPVMYIH RQSAYLLGRH RRIADIPIDH PSCSKQHAVF QYRLVEYTRA DGTVGRRVKP YIIDLGSGNG TFLNNKRIEP QRYYELKEKD VLKFGFSSRE YVLLHESSDT SEIDRKDDED EEEEEEVSDS
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SNIP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 20% glycerol and 100mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers