missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNIP1 Protein
A cDNA sequence encoding the SNIP1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | SNIP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943899
|
enQuireBio™
QP13539-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933899
|
enQuireBio™
QP13539-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923899
|
enQuireBio™
QP13539-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SNIP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SNIP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 20% glycerol and 100mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRWRLYPFKN DEVLPVMYIH RQSAYLLGRH RRIADIPIDH PSCSKQHAVF QYRLVEYTRA DGTVGRRVKP YIIDLGSGNG TFLNNKRIEP QRYYELKEKD VLKFGFSSRE YVLLHESSDT SEIDRKDDED EEEEEEVSDS | |
Greater than 90.0% as determined by SDS-PAGE. |