missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNAPC1 Protein
A cDNA sequence encoding the SNAPC1 was constructed and used to recombinantly synthesize the protein.
£104.00 - £3130.00
Specifications
Name | SNAPC1 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963879
|
enQuireBio™
QP13533-5UG |
5 μg |
£104.00
5µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15953879
|
enQuireBio™
QP13533-20UG |
20 μg |
£159.00
20µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15943879
|
enQuireBio™
QP13533-1MG |
1 mg |
£3130.00
1mg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
SNAPC1 Protein | |
His | |
Human | |
Greater than 80.0% as determined by SDS-PAGE. |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGTPPGL QTDCEALLSR FQETDSVRFE DFTELWRNMK FGTIFCGRMR NLEKNMFTKE ALALAWRYFL PPYTFQIRVG ALYLLYGLYN TQLCQPKQKI RVALKDWDEV LKFQQDLVNA QHFDAAYIFR KLRLDRAFHF TAMPKLLSYR MKKKIHRAEV TEEFKDPSDR VMKLITSDVL EEMLNVHDHY QNMKHVISVD KSKPDKALSL IKDDFFDNIK NIVLEHQQWH KDRKNPSLKS KTNDGEEKME GNSQETERCE RAESLAKIKS KAFSVVIQAS KSRRHRQVKL DSSDSDSASG QGQVKATRKK EKKERLKPAG RKMSLRNKGN VQNIHKEDKP LSLSMPVITE EEENESLSGT EFTASKKRRK H |