missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNAPC1 Protein
A cDNA sequence encoding the SNAPC1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13533-1mg
Additional Details : Weight : 0.01000kg
Specifications
SNAPC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGTPPGL QTDCEALLSR FQETDSVRFE DFTELWRNMK FGTIFCGRMR NLEKNMFTKE ALALAWRYFL PPYTFQIRVG ALYLLYGLYN TQLCQPKQKI RVALKDWDEV LKFQQDLVNA QHFDAAYIFR KLRLDRAFHF TAMPKLLSYR MKKKIHRAEV TEEFKDPSDR VMKLITSDVL EEMLNVHDHY QNMKHVISVD KSKPDKALSL IKDDFFDNIK NIVLEHQQWH KDRKNPSLKS KTNDGEEKME GNSQETERCE RAESLAKIKS KAFSVVIQAS KSRRHRQVKL DSSDSDSASG QGQVKATRKK EKKERLKPAG RKMSLRNKGN VQNIHKEDKP LSLSMPVITE EEENESLSGT EFTASKKRRK H | |
Greater than 80.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNAPC1 protein solution (1 mg/ml) contains 20mM Tris-HCl (pH 8.0) and 10% glycerol. |