missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SKA1 Protein

A cDNA sequence encoding the SKA1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13515-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943819

  • £98.39 / 1µg
Estimated Shipment: 23-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SKA1 Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SKA1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMASSDLE QLCSHVNEKI GNIKKTLSLR NCGQEPTLKT VLNKIGDEII VINELLNKLE LEIQYQEQTN NSLKELCESL EEDYKDIEHL KENVPSHLPQ VTVTQSCVKG SDLDPEEPIK VEEPEPVKKP PKEQRSIKEM PFITCDEFNG VPSYMKSRLT YNQINDVIKE INKAVISKYK ILHQPKKSMN SVTRNLYHRF IDEETKDTKG RYFIVEADIK EFTTLKADKK FHVLLNILRH CRRLSEVRGG GLTRYVIT
Videos
SDS
Documents
Certificates
Special Offers

Special Offers