missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SKA1 Protein
A cDNA sequence encoding the SKA1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | SKA1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943819
|
enQuireBio™
QP13515-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963819
|
enQuireBio™
QP13515-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953819
|
enQuireBio™
QP13515-50UG |
50 μg |
£1320.00
50µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SKA1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SKA1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASSDLE QLCSHVNEKI GNIKKTLSLR NCGQEPTLKT VLNKIGDEII VINELLNKLE LEIQYQEQTN NSLKELCESL EEDYKDIEHL KENVPSHLPQ VTVTQSCVKG SDLDPEEPIK VEEPEPVKKP PKEQRSIKEM PFITCDEFNG VPSYMKSRLT YNQINDVIKE INKAVISKYK ILHQPKKSMN SVTRNLYHRF IDEETKDTKG RYFIVEADIK EFTTLKADKK FHVLLNILRH CRRLSEVRGG GLTRYVIT | |
Greater than 80.0% as determined by SDS-PAGE. |