missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human sFRP-3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£327.21
Specifications
Gene ID (Entrez) | 2487 |
---|---|
Molecular Weight (g/mol) | 33.2 kDa |
Name | sFRP-3 |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived human sFRP-3 protein Ala33-Asn325 |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18178280
|
R&D Systems™
7584-SF-025/CF |
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl. | 25μg |
£327.21
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18308757
|
R&D Systems™
7584-SF-025 |
£327.21
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human sFRP-3 Protein is derived from NS0. The Recombinant Human sFRP-3 Protein has been validated for the following applications: Bioactivity.Specifications
2487 | |
sFRP-3 | |
Mouse myeloma cell line,NS0-derived human sFRP-3 protein Ala33-Asn325 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
33.2 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |