missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human sFRP-3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 7584-SF-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human sFRP-3 Protein is derived from NS0. The Recombinant Human sFRP-3 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein. | |
33.2 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
2487 | |
sFRP-3 | |
Mouse myeloma cell line,NS0-derived human sFRP-3 protein Ala33-Asn325 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |