missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCP2D1 Protein

A cDNA sequence encoding the SCP2D1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13436-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15973559

  • £99.15 / 2µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

140856
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGS MWKRSDH QPKIKAEDGP LVGQFEVLGS VPEPAMPHPL ELSEFESFPV FQDIRLHIRE VGAQLVKKVN AVFQLDITKN GKTILRWTID LKNGSGDMYP GPARLPADTV FTIPESVFME LVLGKMNPQK AFLAGKFKVS GKVLLSWKLE RVFKDWAKF
Greater than 85% as determined by SDS-PAGE.
SCP2D1 Protein
Research Use Only
SCP2D1
Human
His
SCP2D1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers