missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCP2D1 Protein
A cDNA sequence encoding the SCP2D1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 140856 |
---|---|
Name | SCP2D1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | SCP2D1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15973559
|
enQuireBio™
QP13436-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963559
|
enQuireBio™
QP13436-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953559
|
enQuireBio™
QP13436-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
140856 | |
Research Use Only | |
SCP2D1 | |
Human | |
His | |
SCP2D1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
SCP2D1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGS MWKRSDH QPKIKAEDGP LVGQFEVLGS VPEPAMPHPL ELSEFESFPV FQDIRLHIRE VGAQLVKKVN AVFQLDITKN GKTILRWTID LKNGSGDMYP GPARLPADTV FTIPESVFME LVLGKMNPQK AFLAGKFKVS GKVLLSWKLE RVFKDWAKF | |
Greater than 85% as determined by SDS-PAGE. |