missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCG5 Protein

A cDNA sequence encoding the SCG5 was constructed and used to recombinantly synthesize the protein.

£98.39 - £5741.17

Specifications

Name SCG5 Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15973529
View Documents Promotion Details
enQuireBio™
QP13427-2UG
2 μg
£98.39
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15953529
View Documents Promotion Details
enQuireBio™
QP13427-10UG
10 μg
£151.37
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15963529
View Documents Promotion Details
enQuireBio™
QP13427-1MG
1 mg
£5741.17
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

SCG5 Protein
His
Human
Greater than 85% as determined by SDS-PAGE.
Recombinant Protein
E. coli
MYSPRTPDRV SEADIQRLLH GVMEQLGIAR PRVEYPAHQA MNLVGPQSIE GGAHEGLQHL GPFGNIPNIV AELTGDNIPK DFSEDQGYPDPPNPCPVGKT ADDGCLENTP DTAEFSREFQ LHQHLSDPEH DYPGLGKWNK KLLYEKMKGG ERRKRRSVNP YLQGQRLDNV VAKKSVPHFS DEDKDPELEH HHHHH
SDS
Product Certifications