missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCG5 Protein
A cDNA sequence encoding the SCG5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13427-10ug
Additional Details : Weight : 0.01000kg
Specifications
SCG5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MYSPRTPDRV SEADIQRLLH GVMEQLGIAR PRVEYPAHQA MNLVGPQSIE GGAHEGLQHL GPFGNIPNIV AELTGDNIPK DFSEDQGYPDPPNPCPVGKT ADDGCLENTP DTAEFSREFQ LHQHLSDPEH DYPGLGKWNK KLLYEKMKGG ERRKRRSVNP YLQGQRLDNV VAKKSVPHFS DEDKDPELEH HHHHH | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FABP3 His-Tag 0.5 mg/ml protein solution is supplied in 20mM Tris-HCl pH 8.0, 0.1mM PMSF, 1mM DTT, 0.1M NaCl and 20% glycerol. |