missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RTN4IP1 Protein
A cDNA sequence encoding the RTN4IP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13369-20ug
Additional Details : Weight : 0.01000kg
Specifications
RTN4IP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTVMPAWV IDKYGKNEVL RFTQNMMMPI IHYPNEVIVK VHAASVNPID VNMRSGYGAT ALNMKRDPLH VKIKGEEFPL TLGRDVSGVV MECGLDVKYF KPGDEVWAAV PPWKQGTLSE FVVVSGNEVS HKPKSLTHTQ AASLPYVALT AWSAINKVGG LNDKNCTGKR VLILGASGGV GTFAIQVMKA WDAHVTAVCS QDASELVRKL GADDVIDYKS GSVEEQLKSL KPFDFILDNV GGSTETWAPD FLKKWSGATY VTLVTPFLLN MDRLGIADGM LQTGVTVGSK ALKHFWKGVH YRWAFFMASG PCLDDIAELV DAGKIRPVIE QTFPFSKVPE AFLKVERGHA RGKTVINVV | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RTN4IP1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |