missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RTN4IP1 Protein
A cDNA sequence encoding the RTN4IP1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | RTN4IP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15983349
|
enQuireBio™
QP13369-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15973349
|
enQuireBio™
QP13369-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963349
|
enQuireBio™
QP13369-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RTN4IP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RTN4IP1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTVMPAWV IDKYGKNEVL RFTQNMMMPI IHYPNEVIVK VHAASVNPID VNMRSGYGAT ALNMKRDPLH VKIKGEEFPL TLGRDVSGVV MECGLDVKYF KPGDEVWAAV PPWKQGTLSE FVVVSGNEVS HKPKSLTHTQ AASLPYVALT AWSAINKVGG LNDKNCTGKR VLILGASGGV GTFAIQVMKA WDAHVTAVCS QDASELVRKL GADDVIDYKS GSVEEQLKSL KPFDFILDNV GGSTETWAPD FLKKWSGATY VTLVTPFLLN MDRLGIADGM LQTGVTVGSK ALKHFWKGVH YRWAFFMASG PCLDDIAELV DAGKIRPVIE QTFPFSKVPE AFLKVERGHA RGKTVINVV | |
Greater than 95% as determined by SDS-PAGE. |