missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RTN4IP1 Protein

A cDNA sequence encoding the RTN4IP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13369-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963349

  • £2975.00 / 1mg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RTN4IP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSTVMPAWV IDKYGKNEVL RFTQNMMMPI IHYPNEVIVK VHAASVNPID VNMRSGYGAT ALNMKRDPLH VKIKGEEFPL TLGRDVSGVV MECGLDVKYF KPGDEVWAAV PPWKQGTLSE FVVVSGNEVS HKPKSLTHTQ AASLPYVALT AWSAINKVGG LNDKNCTGKR VLILGASGGV GTFAIQVMKA WDAHVTAVCS QDASELVRKL GADDVIDYKS GSVEEQLKSL KPFDFILDNV GGSTETWAPD FLKKWSGATY VTLVTPFLLN MDRLGIADGM LQTGVTVGSK ALKHFWKGVH YRWAFFMASG PCLDDIAELV DAGKIRPVIE QTFPFSKVPE AFLKVERGHA RGKTVINVV
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RTN4IP1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers