missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPS4X Protein

A cDNA sequence encoding the RPS4X was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13358-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933319

  • £2975.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RPS4X Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMARGPKK HLKRVAAPKH WMLDKLTGVF APRPSTGPHK LRECLPLIIF LRNRLKYALT GDEVKKICMQ RFIKIDGKVR TDITYPAGFM DVISIDKTGE NFRLIYDTKG RFAVHRITPE EAKYKLCKVR KIFVGTKGIP HLVTHDARTI RYPDPLIKVN DTIQIDLETG KITDFIKFDT GNLCMVTGGA NLGRIGVITN RERHPGSFDV VHVKDANGNS FATRLSNIFV IGKGNKPWIS LPRGKGIRLT IAEERDKRLA AKQSSG
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RPS4X solution (0.5 mg/ml) contains 20mM Tris-HCl(pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers