missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS4X Protein
A cDNA sequence encoding the RPS4X was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | RPS4X Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953319
|
enQuireBio™
QP13358-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943319
|
enQuireBio™
QP13358-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933319
|
enQuireBio™
QP13358-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RPS4X Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RPS4X solution (0.5 mg/ml) contains 20mM Tris-HCl(pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMARGPKK HLKRVAAPKH WMLDKLTGVF APRPSTGPHK LRECLPLIIF LRNRLKYALT GDEVKKICMQ RFIKIDGKVR TDITYPAGFM DVISIDKTGE NFRLIYDTKG RFAVHRITPE EAKYKLCKVR KIFVGTKGIP HLVTHDARTI RYPDPLIKVN DTIQIDLETG KITDFIKFDT GNLCMVTGGA NLGRIGVITN RERHPGSFDV VHVKDANGNS FATRLSNIFV IGKGNKPWIS LPRGKGIRLT IAEERDKRLA AKQSSG | |
Greater than 95% as determined by SDS-PAGE. |