missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPS27A Protein

A cDNA sequence encoding the RPS27A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13356-B-EC-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963309

  • £880.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

7311; 7314; 6233; 7316
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
The Ubiquitin protein solution contains 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
RPS27A Protein
Research Use Only
RPS27A; UBB; UBC; UBA52
Human
Biotin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers