missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS27A Protein
A cDNA sequence encoding the RPS27A was constructed and used to recombinantly synthesize the protein.
£99.15 - £880.00
Specifications
Gene ID (Entrez) | 7311; 7314; 6233; 7316 |
---|---|
Name | RPS27A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | RPS27A; UBB; UBC; UBA52 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15983309
|
enQuireBio™
QP13356-B-EC-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963309
|
enQuireBio™
QP13356-B-EC-100UG |
100 μg |
£880.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7311; 7314; 6233; 7316 | |
Research Use Only | |
RPS27A; UBB; UBC; UBA52 | |
Human | |
Biotin |
RPS27A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
The Ubiquitin protein solution contains 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |