missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RHEB Protein

A cDNA sequence encoding the RHEB was constructed and used to recombinantly synthesize the protein.

£98.39 - £1989.41

Specifications

Name RHEB Protein
Product Type Recombinant Protein
Protein Tag Untagged
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15963169
View Documents Promotion Details
enQuireBio™
QP13313-10UG
10 μg
£98.39
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15983169
View Documents Promotion Details
enQuireBio™
QP13313-50UG
50 μg
£151.37
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15973169
View Documents Promotion Details
enQuireBio™
QP13313-1MG
1 mg
£1989.41
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

RHEB Protein
Untagged
Human
Greater than 95.0% as determined by SDS-PAGE.
Recombinant Protein
E. coli
MASMTGGQQM GRGSASMPQS KSRKIAILGY RSVGKSSLTI QFVEGQFVDS YDPTIENTFT KLITVNGQEY HLQLVDTAGQ DEYSIFPQTY SIDINGYILV YSVTSIKSFE VIKVIHGKLL DMVGKVQIPI MLVGNKKDLH MERVISYEEG KALAESWNAA FLESSAKENQ TAVDVFRRII LEAEKMDGAA SQGKSSC
SDS
Product Certifications