missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RHEB Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
Specifications
Specifications
| Name | RHEB Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MASMTGGQQM GRGSASMPQS KSRKIAILGY RSVGKSSLTI QFVEGQFVDS YDPTIENTFT KLITVNGQEY HLQLVDTAGQ DEYSIFPQTY SIDINGYILV YSVTSIKSFE VIKVIHGKLL DMVGKVQIPI MLVGNKKDLH MERVISYEEG KALAESWNAA FLESSAKENQ TAVDVFRRII LEAEKMDGAA SQGKSSC |
| Buffer | The RHEB protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction