missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RGS17 Protein
A cDNA sequence encoding the RGS17 was constructed and used to recombinantly synthesize the protein.
£104.00 - £3130.00
Specifications
Name | RGS17 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963159
|
enQuireBio™
QP13309-5UG |
5 μg |
£104.00
5µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15953159
|
enQuireBio™
QP13309-25UG |
25 μg |
£159.00
25µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15943159
|
enQuireBio™
QP13309-1MG |
1 mg |
£3130.00
1mg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
RGS17 Protein | |
His | |
Human | |
Greater than 90% as determined by SDS-PAGE. |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRKRQQSQNE GTPAVSQAPG NQRPNNTCCF CWCCCCSCSC LTVRNEERGE NAGRPTHTTK MESIQVLEEC QNPTAEEVLS WSQNFDKMMK APAGRNLFRE FLRTEYSEEN LLFWLACEDL KKEQNKKVIE EKARMIYEDY ISILSPKEVS LDSRVREVIN RNLLDPNPHM YEDAQLQIYT LMHRDSFPRF LNSQIYKSFV ESTAGSSSES |