missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RGS17 Protein

A cDNA sequence encoding the RGS17 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13309-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963159

  • £99.15 / 5µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RGS17 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MRKRQQSQNE GTPAVSQAPG NQRPNNTCCF CWCCCCSCSC LTVRNEERGE NAGRPTHTTK MESIQVLEEC QNPTAEEVLS WSQNFDKMMK APAGRNLFRE FLRTEYSEEN LLFWLACEDL KKEQNKKVIE EKARMIYEDY ISILSPKEVS LDSRVREVIN RNLLDPNPHM YEDAQLQIYT LMHRDSFPRF LNSQIYKSFV ESTAGSSSES
Greater than 90% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RGS17 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl,1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers